Product Details

SNP ID
rs143205112
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:46834942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTGAGCAGTCATTATCTGTTGT[C/T]TTGGCATCTTGCTTTGAATTCTTTT
Phenotype
MIM: 182135
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HTR2A PubMed Links

Gene Details

Gene
HTR2A
Gene Name
5-hydroxytryptamine receptor 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000621.4 1302 Silent Mutation AAA,AAG K437K NP_000612.1
NM_001165947.2 1302 Silent Mutation AAA,AAG K353K NP_001159419.1

View Full Product Details