Product Details

SNP ID
rs143296005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36433135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGATGCTTGTCAGATACTCACC[A/G]GAGCCCCGCTGGGCCAGGATCCCCC
Phenotype
MIM: 604036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNA1 PubMed Links

Gene Details

Gene
CCNA1
Gene Name
cyclin A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111045.1 217 Missense Mutation AGA,GGA R70G NP_001104515.1
NM_001111046.1 217 Missense Mutation AGA,GGA R27G NP_001104516.1
NM_001111047.1 217 Missense Mutation AGA,GGA R27G NP_001104517.1
NM_003914.3 217 Missense Mutation AGA,GGA R71G NP_003905.1
XM_011535294.2 217 Missense Mutation AGA,GGA R27G XP_011533596.1
XM_011535295.2 217 Missense Mutation AGA,GGA R27G XP_011533597.1
XM_011535296.2 217 Missense Mutation AGA,GGA R27G XP_011533598.1

View Full Product Details