Product Details

SNP ID
rs144632306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99201481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACTGATGAGAGTGCTTTCAAGT[A/G]GAGGGAAGTTAGGAAGTCGTGGCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBAC2 PubMed Links

Gene Details

Gene
UBAC2
Gene Name
UBA domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144072.1 136 Intron NP_001137544.1
NM_177967.3 136 Missense Mutation AGA,GGA R24G NP_808882.1
XM_006719948.3 136 UTR 5 XP_006720011.1
XM_011521082.2 136 Intron XP_011519384.1
XM_011521083.2 136 Missense Mutation AGA,GGA R7G XP_011519385.1
XM_011521084.2 136 Intron XP_011519386.1
XM_017020553.1 136 Intron XP_016876042.1
Gene
UBAC2-AS1
Gene Name
UBAC2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details