Product Details
- SNP ID
-
rs145466956
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:48490114 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAACTTTCCTGCAGATGGGATCA[A/G]TTTTTAAAGGCTCCAACTCTGCTTG
- Phenotype
-
MIM: 614041
MIM: 603524
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RB1
PubMed Links
Gene Details
- Gene
- RB1
- Gene Name
- RB transcriptional corepressor 1
There are no transcripts associated with this gene.
- Gene
- RCBTB2
- Gene Name
- RCC1 and BTB domain containing protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001268.3 |
1921 |
Silent Mutation |
AAC,AAT |
N551N |
NP_001259.1 |
NM_001286830.1 |
1921 |
Silent Mutation |
AAC,AAT |
N556N |
NP_001273759.1 |
NM_001286831.1 |
1921 |
Silent Mutation |
AAC,AAT |
N527N |
NP_001273760.1 |
NM_001286832.1 |
1921 |
Silent Mutation |
AAC,AAT |
N277N |
NP_001273761.1 |
XM_005266242.2 |
1921 |
Silent Mutation |
AAC,AAT |
N551N |
XP_005266299.1 |
XM_005266244.2 |
1921 |
Silent Mutation |
AAC,AAT |
N527N |
XP_005266301.1 |
XM_006719759.3 |
1921 |
Silent Mutation |
AAC,AAT |
N551N |
XP_006719822.1 |
XM_011534894.2 |
1921 |
Silent Mutation |
AAC,AAT |
N556N |
XP_011533196.1 |
XM_011534895.1 |
1921 |
Silent Mutation |
AAC,AAT |
N277N |
XP_011533197.1 |
XM_017020369.1 |
1921 |
Silent Mutation |
AAC,AAT |
N561N |
XP_016875858.1 |
XM_017020370.1 |
1921 |
Silent Mutation |
AAC,AAT |
N561N |
XP_016875859.1 |
XM_017020371.1 |
1921 |
Silent Mutation |
AAC,AAT |
N556N |
XP_016875860.1 |
XM_017020372.1 |
1921 |
Silent Mutation |
AAC,AAT |
N556N |
XP_016875861.1 |
XM_017020373.1 |
1921 |
Silent Mutation |
AAC,AAT |
N555N |
XP_016875862.1 |
XM_017020374.1 |
1921 |
Intron |
|
|
XP_016875863.1 |
XM_017020375.1 |
1921 |
Intron |
|
|
XP_016875864.1 |
XM_017020376.1 |
1921 |
Intron |
|
|
XP_016875865.1 |
XM_017020377.1 |
1921 |
Intron |
|
|
XP_016875866.1 |
XM_017020378.1 |
1921 |
Silent Mutation |
AAC,AAT |
N277N |
XP_016875867.1 |
XM_017020379.1 |
1921 |
Silent Mutation |
AAC,AAT |
N277N |
XP_016875868.1 |
View Full Product Details