Product Details

SNP ID
rs146939104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:36950042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCATTTGTAGCCAGCTCCAGAAT[A/G]GAACTAATTTAGAACCTGTGATTTA
Phenotype
MIM: 604565
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG5 PubMed Links

Gene Details

Gene
ALG5
Gene Name
ALG5, dolichyl-phosphate beta-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142364.1 942 Missense Mutation CCA,CTA P262L NP_001135836.1
NM_013338.4 942 Missense Mutation CCA,CTA P292L NP_037470.1
XM_017020552.1 942 Missense Mutation CCA,CTA P229L XP_016876041.1

View Full Product Details