Product Details

SNP ID
rs147117976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:45702851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATTCCAATCAGTGTGGTTCTAC[C/G]TCAGGTAGGGATAATCACAGAAGGA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01055 PubMed Links

Gene Details

Gene
LINC01055
Gene Name
long intergenic non-protein coding RNA 1055
There are no transcripts associated with this gene.

Gene
SPERT
Gene Name
spermatid associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286341.1 239 Intron NP_001273270.1
NM_001286342.1 239 Intron NP_001273271.1
NM_152719.2 239 Missense Mutation CCT,CGT P51R NP_689932.1
XM_011534971.2 239 Missense Mutation CCT,CGT P51R XP_011533273.1

View Full Product Details