Product Details

SNP ID
rs137933758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77343186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAAAATTAAATTTACCTTCAATC[C/T]CTTCCTCCTCTTCCTCTTCTTCATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMED8 PubMed Links

Gene Details

Gene
TMED8
Gene Name
transmembrane p24 trafficking protein family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_213601.1 651 Missense Mutation GAG,GGG E251G NP_998766.1
XM_017021223.1 651 Missense Mutation GAG,GGG E358G XP_016876712.1
XM_017021224.1 651 Missense Mutation GAG,GGG E188G XP_016876713.1
XM_017021225.1 651 Missense Mutation GAG,GGG E183G XP_016876714.1

View Full Product Details