Product Details

SNP ID
rs138488746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104756628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCCATCTGGGGTAGCGTCCATTG[C/T]CTGTTCCTCATGCGTGCGAGCCGTG
Phenotype
MIM: 612498 MIM: 605567
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADSSL1 PubMed Links

Gene Details

Gene
ADSSL1
Gene Name
adenylosuccinate synthase like 1
There are no transcripts associated with this gene.

Gene
SIVA1
Gene Name
SIVA1 apoptosis inducing factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006427.3 212 Missense Mutation GCC,GTC A113V NP_006418.2
NM_021709.2 212 Missense Mutation GCC,GTC A48V NP_068355.1
XM_011536360.2 212 Missense Mutation GCC,GTC A113V XP_011534662.1

View Full Product Details