Product Details
- SNP ID
-
rs138703292
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:75070711 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGACTCTTCCCAGCAGTCCTTGAA[A/G]GGGCACCTGAGGAACAATTTCCAGA
- Phenotype
-
MIM: 600875
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACYP1
PubMed Links
Gene Details
- Gene
- ACYP1
- Gene Name
- acylphosphatase 1
There are no transcripts associated with this gene.
- Gene
- ZC2HC1C
- Gene Name
- zinc finger C2HC-type containing 1C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042430.2 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
NP_001035895.1 |
NM_024643.3 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
NP_078919.2 |
XM_005268062.3 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_005268119.1 |
XM_005268063.4 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_005268120.1 |
XM_006720255.3 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_006720318.1 |
XM_011537155.2 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_011535457.1 |
XM_011537156.2 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_011535458.1 |
XM_011537157.2 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_011535459.1 |
XM_017021643.1 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_016877132.1 |
XM_017021644.1 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_016877133.1 |
XM_017021645.1 |
266 |
Silent Mutation |
AAA,AAG |
K46K |
XP_016877134.1 |
View Full Product Details