Product Details

SNP ID
rs138723340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94176479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTGATTAGTTTTAAGTTCATT[A/G]TAGAGTAGGAACATTATACGGGATC
Phenotype
MIM: 616790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP4R4 PubMed Links

Gene Details

Gene
PPP4R4
Gene Name
protein phosphatase 4 regulatory subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020958.2 Intron NP_066009.2
NM_058237.1 Intron NP_478144.1
XM_005267930.2 Intron XP_005267987.1
XM_011537038.2 Intron XP_011535340.1
XM_011537039.2 Intron XP_011535341.1
XM_011537040.2 Intron XP_011535342.1
XM_017021528.1 Intron XP_016877017.1
XM_017021529.1 Intron XP_016877018.1
XM_017021530.1 Intron XP_016877019.1

View Full Product Details