Product Details

SNP ID
rs139231018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74763951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTGAATCTCCCCCTGTGCCGCC[A/T]GGGTCCTATATGCCCCCATCTCAGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
YLPM1 PubMed Links

Gene Details

Gene
YLPM1
Gene Name
YLP motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019589.2 630 Silent Mutation CCA,CCT P154P NP_062535.2
XM_005267860.3 630 Silent Mutation CCA,CCT P154P XP_005267917.1
XM_011536966.2 630 Intron XP_011535268.1
XM_011536967.1 630 Silent Mutation CCA,CCT P154P XP_011535269.1

View Full Product Details