Product Details

SNP ID
rs139267307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77275773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGTCAAAAGGGCTGTAGCAGTG[A/G]GAAAGGGAGGTCATCGGGCCCCAGA
Phenotype
MIM: 605304 MIM: 607439
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1260A PubMed Links

Gene Details

Gene
MIR1260A
Gene Name
microRNA 1260a
There are no transcripts associated with this gene.

Gene
NGB
Gene Name
neuroglobin
There are no transcripts associated with this gene.

Gene
POMT2
Gene Name
protein O-mannosyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013382.5 4159 UTR 3 NP_037514.2
XM_011536675.2 4159 UTR 3 XP_011534977.1
XM_011536676.2 4159 UTR 3 XP_011534978.1
XM_011536677.2 4159 UTR 3 XP_011534979.1
XM_011536679.2 4159 UTR 3 XP_011534981.1
XM_017021244.1 4159 UTR 3 XP_016876733.1
XM_017021245.1 4159 UTR 3 XP_016876734.1

View Full Product Details