Product Details

SNP ID
rs139735871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49577819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGGAGGAGATGGGTGTCACCTC[C/T]ATAGGCAGTGCCAAGGAAGACAGCT
Phenotype
MIM: 612177 MIM: 603633
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RN7SL1 PubMed Links

Gene Details

Gene
RN7SL1
Gene Name
RNA, 7SL, cytoplasmic 1
There are no transcripts associated with this gene.

Gene
RPS29
Gene Name
ribosomal protein S29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030001.2 267 Nonsense Mutation NP_001025172.1
NM_001032.4 267 Intron NP_001023.1

View Full Product Details