Product Details

SNP ID
rs140020925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20060510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCATGGATAATTGGTTTTTTACAC[A/T]CCATAAGCCAGATAGTTTTAACAAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4L1 PubMed Links

Gene Details

Gene
OR4L1
Gene Name
olfactory receptor family 4 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004717.1 466 Missense Mutation ACC,TCC T156S NP_001004717.1

View Full Product Details