Product Details
- SNP ID
-
rs140087770
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:93908017 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCAGAACTGGGACCTGAAGCCAG[G/T]TTTATGGCTCCAGAGCTCCCCAGTC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FAM181A-AS1
PubMed Links
Gene Details
- Gene
- FAM181A-AS1
- Gene Name
- FAM181A antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details