Product Details

SNP ID
rs140646891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19921120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAACCTGCCTTTTTGTGGACCT[A/G]ATGTAGTAGACAGCTTTTTTTGTGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K1 PubMed Links

Gene Details

Gene
OR4K1
Gene Name
olfactory receptor family 4 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004063.2 514 Intron NP_001004063.2
XM_011537153.2 514 Intron XP_011535455.1
Gene
OR4K5
Gene Name
olfactory receptor family 4 subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005483.1 514 Missense Mutation AAT,GAT N172D NP_001005483.1

View Full Product Details