Product Details

SNP ID
rs140724238
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76790616 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGATCCATCAGGCTTACCAGTT[A/C]TGTTCCCATTCTCACAGGACTCAGC
Phenotype
MIM: 609011
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ANGEL1 PubMed Links

Gene Details

Gene
ANGEL1
Gene Name
angel homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015305.3 1960 Missense Mutation AGA,ATA R616I NP_056120.2
XM_017021115.1 1960 UTR 3 XP_016876604.1
Gene
LOC100506603
Gene Name
uncharacterized LOC100506603
There are no transcripts associated with this gene.

Gene
VASH1
Gene Name
vasohibin 1
There are no transcripts associated with this gene.

View Full Product Details