Product Details

SNP ID
rs140905741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:101979431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTACGAAACTTTGCATTCTTTC[A/G]TTAGCAATGCAGTGGCTCCTTTTTT
Phenotype
MIM: 600112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DYNC1H1 PubMed Links

Gene Details

Gene
DYNC1H1
Gene Name
dynein cytoplasmic 1 heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001376.4 621 Missense Mutation ATT,GTT I153V NP_001367.2

View Full Product Details