Product Details

SNP ID
rs141168105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76788790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGACACATACATGATATGCATAT[C/T]TCACATATGCATGCCTGGCAAGAGG
Phenotype
MIM: 609011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANGEL1 PubMed Links

Gene Details

Gene
ANGEL1
Gene Name
angel homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015305.3 2564 UTR 3 NP_056120.2
XM_017021115.1 2564 Intron XP_016876604.1
Gene
LOC100506603
Gene Name
uncharacterized LOC100506603
There are no transcripts associated with this gene.

Gene
VASH1
Gene Name
vasohibin 1
There are no transcripts associated with this gene.

View Full Product Details