Product Details

SNP ID
rs141657598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49710281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAGCTTTATTTTGTTAATTTA[C/T]GAACAAGAGATGAAACCTACATTTG
Phenotype
MIM: 611281
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHDC1 PubMed Links

Gene Details

Gene
KLHDC1
Gene Name
kelch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172193.2 394 Nonsense Mutation CGA,TGA R102* NP_751943.1
XM_011536422.2 394 Nonsense Mutation CGA,TGA R102* XP_011534724.1

View Full Product Details