Product Details

SNP ID
rs142176719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21058153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCACCTGCCAGACCCCCAACAT[A/G]GCCTGCAAGAATAGCTGTAAAAACT
Phenotype
MIM: 610018 MIM: 605272 MIM: 612485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 332 Intron NP_001269140.1
NM_001282212.1 332 Intron NP_001269141.1
NM_001282213.1 332 Intron NP_001269142.1
NM_001282214.1 332 Intron NP_001269143.1
NM_001282215.1 332 Intron NP_001269144.1
NM_001282216.1 332 Intron NP_001269145.1
NM_001320329.1 332 Intron NP_001307258.1
NM_016250.2 332 Intron NP_057334.1
NM_201535.1 332 Intron NP_963293.1
NM_201536.1 332 Intron NP_963294.1
NM_201537.1 332 Intron NP_963831.1
NM_201538.1 332 Intron NP_963832.1
NM_201539.1 332 Intron NP_963833.1
NM_201540.1 332 Intron NP_963834.1
NM_201541.1 332 Intron NP_963835.1
XM_011536996.2 332 Intron XP_011535298.1
XM_011536997.1 332 Intron XP_011535299.1
XM_011536998.1 332 Intron XP_011535300.1
XM_011536999.1 332 Intron XP_011535301.1
XM_011537001.1 332 Intron XP_011535303.1
XM_011537002.1 332 Intron XP_011535304.1
XM_017021480.1 332 Intron XP_016876969.1
XM_017021481.1 332 Intron XP_016876970.1
XM_017021482.1 332 Intron XP_016876971.1
XM_017021483.1 332 Intron XP_016876972.1
XM_017021484.1 332 Intron XP_016876973.1
XM_017021485.1 332 Intron XP_016876974.1
XM_017021486.1 332 Intron XP_016876975.1
XM_017021487.1 332 Intron XP_016876976.1
XM_017021488.1 332 Intron XP_016876977.1
XM_017021489.1 332 Intron XP_016876978.1
XM_017021490.1 332 Intron XP_016876979.1
XM_017021491.1 332 Intron XP_016876980.1
XM_017021492.1 332 Intron XP_016876981.1
XM_017021493.1 332 Intron XP_016876982.1
XM_017021494.1 332 Intron XP_016876983.1
XM_017021495.1 332 Intron XP_016876984.1
XM_017021496.1 332 Intron XP_016876985.1
XM_017021497.1 332 Intron XP_016876986.1
Gene
RNASE8
Gene Name
ribonuclease A family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138331.2 332 Missense Mutation ATA,ATG I87M NP_612204.1

View Full Product Details