Product Details

SNP ID
rs142684401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103556804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAAAAGCTGTGCTCAAAAAAAA[A/T]ATAAAAAAACCCAAAACCTTGCATG
Phenotype
MIM: 616003 MIM: 603885
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APOPT1 PubMed Links

Gene Details

Gene
APOPT1
Gene Name
apoptogenic 1, mitochondrial
There are no transcripts associated with this gene.

Gene
BAG5
Gene Name
BCL2 associated athanogene 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015048.2 4795 UTR 3 NP_001015048.1
NM_001015049.2 4795 UTR 3 NP_001015049.1
NM_004873.3 4795 UTR 3 NP_004864.1

View Full Product Details