Product Details

SNP ID
rs142685746
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105011427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGGGGTTTTTAGTCTCCAGC[A/G]TTTCAAATATCTGATCAAGTGACTT
Phenotype
MIM: 612270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDCA4 PubMed Links

Gene Details

Gene
CDCA4
Gene Name
cell division cycle associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017955.3 599 Missense Mutation ACG,ATG T168M NP_060425.2
NM_145701.2 599 Missense Mutation ACG,ATG T168M NP_663747.1

View Full Product Details