Product Details

SNP ID
rs143404451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39240565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTTCTTCATTTTGACAGAAAG[A/G]AAAGGAGTTTGGATATTTTCCCAGA
Phenotype
MIM: 608001
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIA2 PubMed Links

Gene Details

Gene
MIA2
Gene Name
melanoma inhibitory activity 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054024.3 448 Missense Mutation GAA,GGA E85G NP_473365.3

View Full Product Details