Product Details

SNP ID
rs145110813
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20447203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCACGGGGTCCTTTGGGTTCC[A/G]ATTAAGGAACTATCTACTCTGATTC
Phenotype
MIM: 107748 MIM: 610107
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
There are no transcripts associated with this gene.

Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017807.3 1470 UTR 3 NP_060277.1

View Full Product Details