Product Details

SNP ID
rs146168332
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20511099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTATAACCTGTAATGACATGAAG[C/T]GCCAGTTACCAACTGGACAATGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASE10 PubMed Links

Gene Details

Gene
RNASE10
Gene Name
ribonuclease A family member 10 (inactive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012975.1 628 Missense Mutation CGC,TGC R210C NP_001012993.1

View Full Product Details