Product Details

SNP ID
rs146230546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94583415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGATGGGCTGGAGTGGAACAG[A/G]AAGTCAAGATTACTTCTTCTGGGAA
Phenotype
MIM: 601841
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA5 PubMed Links

Gene Details

Gene
SERPINA5
Gene Name
serpin family A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000624.5 Intron NP_000615.3

View Full Product Details