Product Details

SNP ID
rs146726398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20447956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGGAGCCACAATGTGCCATGGC[C/T]CGCTCTGTGATCTCTACCAGCATTG
Phenotype
MIM: 107748 MIM: 610107 MIM: 609865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APEX1 PubMed Links

Gene Details

Gene
APEX1
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 1
There are no transcripts associated with this gene.

Gene
OSGEP
Gene Name
O-sialoglycoprotein endopeptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017807.3 1166 Silent Mutation CGA,CGG R247R NP_060277.1
Gene
TMEM55B
Gene Name
transmembrane protein 55B
There are no transcripts associated with this gene.

View Full Product Details