Product Details

SNP ID
rs147004133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74064870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTGAAGGAGGATCGAGAGCC[A/G]GTGAATGAGAACATTGGCAAAGGCA
Phenotype
MIM: 603178
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH6A1 PubMed Links

Gene Details

Gene
ALDH6A1
Gene Name
aldehyde dehydrogenase 6 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278593.1 1442 Silent Mutation NP_001265522.1
NM_001278594.1 1442 Silent Mutation NP_001265523.1
NM_005589.3 1442 Silent Mutation NP_005580.1
XM_017021331.1 1442 Silent Mutation XP_016876820.1
Gene
BBOF1
Gene Name
basal body orientation factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025057.2 1442 UTR 3 NP_079333.2
XM_005268092.3 1442 Intron XP_005268149.1
XM_005268097.3 1442 Intron XP_005268154.1
XM_005268098.4 1442 Intron XP_005268155.1
XM_011537170.2 1442 Intron XP_011535472.1
XM_011537171.2 1442 UTR 3 XP_011535473.1
XM_011537172.1 1442 Intron XP_011535474.1
XM_011537174.2 1442 UTR 3 XP_011535476.1
XM_011537175.2 1442 Intron XP_011535477.1
XM_011537176.2 1442 Intron XP_011535478.1
XM_011537177.2 1442 Intron XP_011535479.1
XM_011537178.2 1442 Intron XP_011535480.1
XM_011537179.2 1442 Intron XP_011535481.1
XM_017021659.1 1442 Intron XP_016877148.1
XM_017021660.1 1442 UTR 3 XP_016877149.1
XM_017021661.1 1442 UTR 3 XP_016877150.1
XM_017021662.1 1442 UTR 3 XP_016877151.1
XM_017021663.1 1442 UTR 3 XP_016877152.1

View Full Product Details