Product Details
- SNP ID
-
rs147216742
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:23561558 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTGCTTCTGAAAGCTGGGCTGCT[G/T]CTTTGCTTTCCTTTGCTTCCTCATC
- Phenotype
-
MIM: 603534
MIM: 611612
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AP1G2
PubMed Links
Gene Details
- Gene
- AP1G2
- Gene Name
- adaptor related protein complex 1 gamma 2 subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282474.1 |
1783 |
Missense Mutation |
GAA,GCA |
E223A |
NP_001269403.1 |
NM_001282475.1 |
1783 |
Missense Mutation |
GAA,GCA |
E532A |
NP_001269404.1 |
NM_003917.4 |
1783 |
Missense Mutation |
GAA,GCA |
E604A |
NP_003908.1 |
XM_005268167.3 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_005268224.1 |
XM_005268168.4 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_005268225.1 |
XM_005268169.3 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_005268226.1 |
XM_005268170.3 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_005268227.1 |
XM_005268172.3 |
1783 |
Missense Mutation |
GAA,GCA |
E604A |
XP_005268229.1 |
XM_005268173.3 |
1783 |
Missense Mutation |
GAA,GCA |
E604A |
XP_005268230.1 |
XM_005268174.3 |
1783 |
Missense Mutation |
GAA,GCA |
E532A |
XP_005268231.1 |
XM_005268175.3 |
1783 |
Missense Mutation |
GAA,GCA |
E532A |
XP_005268232.1 |
XM_005268177.3 |
1783 |
Missense Mutation |
GAA,GCA |
E475A |
XP_005268234.1 |
XM_005268178.3 |
1783 |
Missense Mutation |
GAA,GCA |
E475A |
XP_005268235.1 |
XM_005268179.3 |
1783 |
Missense Mutation |
GAA,GCA |
E475A |
XP_005268236.1 |
XM_005268180.4 |
1783 |
Missense Mutation |
GAA,GCA |
E459A |
XP_005268237.1 |
XM_005268182.3 |
1783 |
Missense Mutation |
GAA,GCA |
E394A |
XP_005268239.1 |
XM_006720301.3 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_006720364.1 |
XM_011537283.2 |
1783 |
Missense Mutation |
GAA,GCA |
E626A |
XP_011535585.1 |
XM_011537286.2 |
1783 |
Missense Mutation |
GAA,GCA |
E223A |
XP_011535588.1 |
XM_017021740.1 |
1783 |
Missense Mutation |
GAA,GCA |
E661A |
XP_016877229.1 |
XM_017021741.1 |
1783 |
Missense Mutation |
GAA,GCA |
E614A |
XP_016877230.1 |
XM_017021742.1 |
1783 |
Missense Mutation |
GAA,GCA |
E532A |
XP_016877231.1 |
XM_017021743.1 |
1783 |
Intron |
|
|
XP_016877232.1 |
XM_017021744.1 |
1783 |
Intron |
|
|
XP_016877233.1 |
XM_017021745.1 |
1783 |
Intron |
|
|
XP_016877234.1 |
XM_017021746.1 |
1783 |
Missense Mutation |
GAA,GCA |
E223A |
XP_016877235.1 |
XM_017021747.1 |
1783 |
Missense Mutation |
GAA,GCA |
E223A |
XP_016877236.1 |
XM_017021748.1 |
1783 |
Missense Mutation |
GAA,GCA |
E223A |
XP_016877237.1 |
- Gene
- JPH4
- Gene Name
- junctophilin 4
There are no transcripts associated with this gene.
- Gene
- LOC102724814
- Gene Name
- uncharacterized LOC102724814
There are no transcripts associated with this gene.
- Gene
- THTPA
- Gene Name
- thiamine triphosphatase
There are no transcripts associated with this gene.
- Gene
- ZFHX2
- Gene Name
- zinc finger homeobox 2
There are no transcripts associated with this gene.
View Full Product Details