Product Details

SNP ID
rs147279776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74085066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTGGGAACATCCACTCTGTCTCT[G/T]CCCCTCGAACATACATTTCTTTCTT
Phenotype
MIM: 603178
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH6A1 PubMed Links

Gene Details

Gene
ALDH6A1
Gene Name
aldehyde dehydrogenase 6 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278593.1 Intron NP_001265522.1
NM_001278594.1 Intron NP_001265523.1
NM_005589.3 Intron NP_005580.1
XM_017021331.1 Intron XP_016876820.1
Gene
BBOF1
Gene Name
basal body orientation factor 1
There are no transcripts associated with this gene.

Gene
LIN52
Gene Name
lin-52 DREAM MuvB core complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024674.2 Intron NP_001019845.1
XM_011537320.2 Intron XP_011535622.1
XM_011537321.2 Intron XP_011535623.1
XM_011537322.2 Intron XP_011535624.2
XM_017021763.1 Intron XP_016877252.1
XM_017021764.1 Intron XP_016877253.1

View Full Product Details