Product Details

SNP ID
rs148657945
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21058000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCAAGGACATGACATCATCTCA[A/G]TGGTTTAAAACTCAGCATGTGCAGC
Phenotype
MIM: 610018 MIM: 605272 MIM: 612485
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links
Additional Information
For this assay, SNP(s) [rs12435011] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 179 Intron NP_001269140.1
NM_001282212.1 179 Intron NP_001269141.1
NM_001282213.1 179 Intron NP_001269142.1
NM_001282214.1 179 Intron NP_001269143.1
NM_001282215.1 179 Intron NP_001269144.1
NM_001282216.1 179 Intron NP_001269145.1
NM_001320329.1 179 Intron NP_001307258.1
NM_016250.2 179 Intron NP_057334.1
NM_201535.1 179 Intron NP_963293.1
NM_201536.1 179 Intron NP_963294.1
NM_201537.1 179 Intron NP_963831.1
NM_201538.1 179 Intron NP_963832.1
NM_201539.1 179 Intron NP_963833.1
NM_201540.1 179 Intron NP_963834.1
NM_201541.1 179 Intron NP_963835.1
XM_011536996.2 179 Intron XP_011535298.1
XM_011536997.1 179 Intron XP_011535299.1
XM_011536998.1 179 Intron XP_011535300.1
XM_011536999.1 179 Intron XP_011535301.1
XM_011537001.1 179 Intron XP_011535303.1
XM_011537002.1 179 Intron XP_011535304.1
XM_017021480.1 179 Intron XP_016876969.1
XM_017021481.1 179 Intron XP_016876970.1
XM_017021482.1 179 Intron XP_016876971.1
XM_017021483.1 179 Intron XP_016876972.1
XM_017021484.1 179 Intron XP_016876973.1
XM_017021485.1 179 Intron XP_016876974.1
XM_017021486.1 179 Intron XP_016876975.1
XM_017021487.1 179 Intron XP_016876976.1
XM_017021488.1 179 Intron XP_016876977.1
XM_017021489.1 179 Intron XP_016876978.1
XM_017021490.1 179 Intron XP_016876979.1
XM_017021491.1 179 Intron XP_016876980.1
XM_017021492.1 179 Intron XP_016876981.1
XM_017021493.1 179 Intron XP_016876982.1
XM_017021494.1 179 Intron XP_016876983.1
XM_017021495.1 179 Intron XP_016876984.1
XM_017021496.1 179 Intron XP_016876985.1
XM_017021497.1 179 Intron XP_016876986.1
Gene
RNASE8
Gene Name
ribonuclease A family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138331.2 179 Silent Mutation CAA,CAG Q36Q NP_612204.1

View Full Product Details