Product Details

SNP ID
rs150338494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49586017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCACATATTGAGGCCATATTTC[C/G]GGATCAGACCGTGCCGGTTTGAACA
Phenotype
MIM: 612177 MIM: 603633
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RN7SL1 PubMed Links

Gene Details

Gene
RN7SL1
Gene Name
RNA, 7SL, cytoplasmic 1
There are no transcripts associated with this gene.

Gene
RPS29
Gene Name
ribosomal protein S29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030001.2 165 Missense Mutation NP_001025172.1
NM_001032.4 165 Intron NP_001023.1

View Full Product Details