Product Details

SNP ID
rs150752116
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95417888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAAGGAGCGGGCGAAGTTGTTGG[A/C]CAGGGTGCAGCTGCGGTCCTCTTCC
Phenotype
MIM: 610861
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00341 PubMed Links

Gene Details

Gene
LINC00341
Gene Name
long intergenic non-protein coding RNA 341
There are no transcripts associated with this gene.

Gene
SYNE3
Gene Name
spectrin repeat containing nuclear envelope family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152592.3 2865 Missense Mutation GCC,TCC A956S NP_689805.3
XM_005267377.3 2865 Missense Mutation GCC,TCC A956S XP_005267434.1
XM_006720063.3 2865 Missense Mutation GCC,TCC A956S XP_006720126.1
XM_011536515.2 2865 Missense Mutation GCC,TCC A964S XP_011534817.1
XM_017021056.1 2865 Missense Mutation GCC,TCC A951S XP_016876545.1
XM_017021057.1 2865 Intron XP_016876546.1

View Full Product Details