Product Details

SNP ID
rs151262490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24607288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGCAGTGTGGTTGCTAAAGTG[C/T]TCATTGAGACATAACCCCAGCCAGC
Phenotype
MIM: 116831
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZMH PubMed Links

Gene Details

Gene
GZMH
Gene Name
granzyme H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270780.1 565 Intron NP_001257709.1
NM_001270781.1 565 Intron NP_001257710.1
NM_033423.4 565 Missense Mutation AAC,AGC N153S NP_219491.1
XM_011536683.2 565 Missense Mutation AAC,AGC N119S XP_011534985.1

View Full Product Details