Product Details
- SNP ID
-
rs111645334
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:76938977 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCCAGAGTTCGAGAACAACTGGG[A/C]AATGTGGCAAGACCCCGTCCCTATG
- Phenotype
-
MIM: 602584
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RCN2
PubMed Links
Gene Details
- Gene
- RCN2
- Gene Name
- reticulocalbin 2
View Full Product Details