Product Details

SNP ID
rs111952796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52193971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCATTTCCAGAGCATGTGGAGA[A/G]GGGGTAAAAGGAAATGTGACTTGAA
Phenotype
MIM: 604447 MIM: 610022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNB5 PubMed Links

Gene Details

Gene
GNB5
Gene Name
G protein subunit beta 5
There are no transcripts associated with this gene.

Gene
LOC100129973
Gene Name
uncharacterized LOC100129973
There are no transcripts associated with this gene.

Gene
MYO5C
Gene Name
myosin VC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018728.3 5322 Silent Mutation NP_061198.2
XM_011521781.2 5322 Silent Mutation XP_011520083.1
XM_017022408.1 5322 Silent Mutation XP_016877897.1
XM_017022409.1 5322 Silent Mutation XP_016877898.1
XM_017022410.1 5322 Silent Mutation XP_016877899.1

View Full Product Details