Product Details

SNP ID
rs112099437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73998396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAGAGCTGCGCAACCAGTCGGT[A/G]CGTGAGTTCCTGGACGGCACCCGCA
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002675.3 662 Silent Mutation GTA,GTG V174V NP_002666.1
NM_033238.2 662 Silent Mutation GTA,GTG V174V NP_150241.2
NM_033239.2 662 Silent Mutation GTA,GTG V174V NP_150242.1
NM_033240.2 662 Silent Mutation GTA,GTG V174V NP_150243.2
NM_033244.3 662 Silent Mutation GTA,GTG V174V NP_150247.2
NM_033246.2 662 Silent Mutation GTA,GTG V174V NP_150249.1
NM_033247.2 662 Silent Mutation GTA,GTG V174V NP_150250.2
NM_033249.2 662 Silent Mutation GTA,GTG V174V NP_150252.1
NM_033250.2 662 Silent Mutation GTA,GTG V174V NP_150253.2
Gene
STOML1
Gene Name
stomatin like 1
There are no transcripts associated with this gene.

View Full Product Details