Product Details

SNP ID
rs112182809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:92796479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTTATGCCTAGAAATCAGAAG[G/T]GCTTTGGAATCCAGCAGCATCATAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ASB9P1 PubMed Links

Gene Details

Gene
ASB9P1
Gene Name
ankyrin repeat and SOCS box containing 9 pseudogene 1
There are no transcripts associated with this gene.

Gene
LOC105370978
Gene Name
uncharacterized LOC105370978
There are no transcripts associated with this gene.

View Full Product Details