Product Details

SNP ID
rs112583776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45366183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCTGGAGCAAGATGCCTACGCA[C/T]CCATTCAATGCCTAGGTAGTTTGTA
Phenotype
MIM: 602360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 1170 Missense Mutation ATG,GTG M152V NP_001307944.1
NM_001482.2 1170 Missense Mutation ATG,GTG M281V NP_001473.1

View Full Product Details