Product Details

SNP ID
rs112867154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50182601 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGGCGCGCACCATCCTGCGGGC[A/G]TTCCTGGAGAAAGCGCGCCAGACGC
Phenotype
MIM: 609123 MIM: 603247
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8B4 PubMed Links

Gene Details

Gene
ATP8B4
Gene Name
ATPase phospholipid transporting 8B4 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024837.3 406 Intron NP_079113.2
XM_011522046.2 406 Intron XP_011520348.1
XM_011522047.2 406 Intron XP_011520349.1
XM_011522048.1 406 Intron XP_011520350.1
XM_011522049.2 406 Intron XP_011520351.1
XM_011522051.2 406 Intron XP_011520353.1
XM_011522052.2 406 Intron XP_011520354.1
XM_011522053.1 406 Intron XP_011520355.1
XM_011522056.2 406 Intron XP_011520358.2
XM_011522058.2 406 Intron XP_011520360.1
XM_011522059.1 406 Intron XP_011520361.1
XM_011522060.1 406 Intron XP_011520362.1
XM_011522061.1 406 Intron XP_011520363.1
XM_011522062.1 406 Intron XP_011520364.1
XM_011522063.1 406 Intron XP_011520365.1
XM_011522064.1 406 Intron XP_011520366.1
XM_011522069.2 406 Intron XP_011520371.1
XM_011522070.1 406 Intron XP_011520372.1
XM_017022587.1 406 Intron XP_016878076.1
XM_017022588.1 406 Intron XP_016878077.1
XM_017022589.1 406 Intron XP_016878078.1
XM_017022590.1 406 Intron XP_016878079.1
XM_017022591.1 406 Intron XP_016878080.1
XM_017022592.1 406 Intron XP_016878081.1
XM_017022593.1 406 Intron XP_016878082.1
XM_017022594.1 406 Intron XP_016878083.1
XM_017022595.1 406 Intron XP_016878084.1
XM_017022596.1 406 Intron XP_016878085.1
XM_017022597.1 406 Intron XP_016878086.1
XM_017022598.1 406 Intron XP_016878087.1
XM_017022599.1 406 Intron XP_016878088.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159629.1 406 Silent Mutation GCA,GCG A58A NP_001153101.1
NM_003645.3 406 Silent Mutation GCA,GCG A58A NP_003636.2

View Full Product Details