Product Details

SNP ID
rs114231786
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40164131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGAATTGTGACTAGTAAACTG[A/T]GTTTGTTTGCTTCTGCTTTTAGCTA
Phenotype
MIM: 602860
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BUB1B PubMed Links

Gene Details

Gene
BUB1B
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001211.5 Intron NP_001202.4

View Full Product Details