Product Details

SNP ID
rs114383018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99972410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAGTGGGCATATCTGGAGGCCC[C/T]GGGGCCCATGCAGGAGACCAAGCTA
Phenotype
MIM: 607511
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS17 PubMed Links

Gene Details

Gene
ADAMTS17
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139057.3 6585 UTR 3 NP_620688.2
XM_005254872.3 6585 UTR 3 XP_005254929.1
XM_011521312.2 6585 UTR 3 XP_011519614.1
XM_017021973.1 6585 UTR 3 XP_016877462.1
XM_017021974.1 6585 Intron XP_016877463.1
XM_017021975.1 6585 UTR 3 XP_016877464.1
XM_017021976.1 6585 UTR 3 XP_016877465.1
XM_017021977.1 6585 Intron XP_016877466.1
XM_017021978.1 6585 UTR 3 XP_016877467.1
XM_017021979.1 6585 UTR 3 XP_016877468.1
XM_017021980.1 6585 UTR 3 XP_016877469.1
XM_017021981.1 6585 Intron XP_016877470.1
XM_017021982.1 6585 Intron XP_016877471.1
XM_017021983.1 6585 Intron XP_016877472.1
XM_017021984.1 6585 Intron XP_016877473.1
Gene
LOC105371022
Gene Name
uncharacterized LOC105371022
There are no transcripts associated with this gene.

View Full Product Details