Product Details
- SNP ID
-
rs114629863
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:23686955 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCGGGGCTGGCGGTGCCGGGCCC[A/G]GCTGGGCCGCGCTCGGGGCCTGGTG
- Phenotype
-
MIM: 602117
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NDN
PubMed Links
Gene Details
- Gene
- NDN
- Gene Name
- necdin, MAGE family member
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002487.2 |
349 |
Missense Mutation |
CCG,CTG |
P88L |
NP_002478.1 |
View Full Product Details