Product Details

SNP ID
rs114860382
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73984590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCAGGGGGCAGTCTACGAGAGC[A/G]CCACTGGGGTAGGTAACAAGAAACC
Phenotype
MIM: 608326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STOML1 PubMed Links

Gene Details

Gene
STOML1
Gene Name
stomatin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256672.1 Intron NP_001243601.1
NM_001256673.1 Intron NP_001243602.1
NM_001256674.1 Intron NP_001243603.1
NM_001256675.1 Intron NP_001243604.1
NM_001256676.1 Intron NP_001243605.1
NM_001256677.1 Intron NP_001243606.1
NM_001324226.1 Intron NP_001311155.1
NM_001324227.1 Intron NP_001311156.1
NM_001324228.1 Intron NP_001311157.1
NM_001324229.1 Intron NP_001311158.1
NM_001324230.1 Intron NP_001311159.1
NM_004809.4 Intron NP_004800.2
XM_005254792.4 Intron XP_005254849.1
XM_006720773.3 Intron XP_006720836.1

View Full Product Details