Product Details
- SNP ID
-
rs137918449
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:28113185 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTGTTGGAGAAGGACTCCATCAC[C/T]TCCCAGAACCACTGGATCAGCGATG
- Phenotype
-
MIM: 605837
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HERC2
PubMed Links
Gene Details
- Gene
- HERC2
- Gene Name
- HECT and RLD domain containing E3 ubiquitin protein ligase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004667.5 |
7556 |
Silent Mutation |
GAA,GAG |
E4706E |
NP_004658.3 |
XM_005268276.4 |
7556 |
Silent Mutation |
GAA,GAG |
E4668E |
XP_005268333.1 |
XM_006720726.3 |
7556 |
Silent Mutation |
GAA,GAG |
E4701E |
XP_006720789.1 |
XM_006720727.3 |
7556 |
Silent Mutation |
GAA,GAG |
E4620E |
XP_006720790.1 |
XM_011522131.2 |
7556 |
Silent Mutation |
GAA,GAG |
E4545E |
XP_011520433.1 |
XM_011522133.2 |
7556 |
Silent Mutation |
GAA,GAG |
E3621E |
XP_011520435.1 |
XM_017022695.1 |
7556 |
Silent Mutation |
GAA,GAG |
E4668E |
XP_016878184.1 |
XM_017022696.1 |
7556 |
Silent Mutation |
GAA,GAG |
E4668E |
XP_016878185.1 |
XM_017022697.1 |
7556 |
Silent Mutation |
GAA,GAG |
E2428E |
XP_016878186.1 |
XM_017022698.1 |
7556 |
Silent Mutation |
GAA,GAG |
E2428E |
XP_016878187.1 |
View Full Product Details