Product Details

SNP ID
rs138467583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:70882560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGAATCACTGGAGTAAATGTCA[A/G]AGAGAGAGGAAGTTCTAGACCACAA
Phenotype
MIM: 612538
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC49 PubMed Links

Gene Details

Gene
LRRC49
Gene Name
leucine rich repeat containing 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199017.2 829 Intron NP_001185946.1
NM_001199018.2 829 Intron NP_001185947.1
NM_001284357.1 829 Intron NP_001271286.1
NM_017691.4 829 Intron NP_060161.2
XM_005254492.4 829 Intron XP_005254549.1
XM_011521715.2 829 Intron XP_011520017.1
XM_011521717.2 829 Intron XP_011520019.1
XM_011521718.2 829 Intron XP_011520020.1
XM_017022357.1 829 Intron XP_016877846.1
XM_017022358.1 829 Intron XP_016877847.1
XM_017022359.1 829 Intron XP_016877848.1
Gene
THAP10
Gene Name
THAP domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020147.3 829 Missense Mutation TCT,TTT S223F NP_064532.1

View Full Product Details