Product Details

SNP ID
rs138507064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73985028 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTTCCTCCTCTACCAGAAAGGT[A/G]CCCCTGAGTTTTCAGCCTAAACTGC
Phenotype
MIM: 102578 MIM: 608326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PML PubMed Links

Gene Details

Gene
PML
Gene Name
promyelocytic leukemia
There are no transcripts associated with this gene.

Gene
STOML1
Gene Name
stomatin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256672.1 Intron NP_001243601.1
NM_001256673.1 Intron NP_001243602.1
NM_001256674.1 Intron NP_001243603.1
NM_001256675.1 Intron NP_001243604.1
NM_001256676.1 Intron NP_001243605.1
NM_001256677.1 Intron NP_001243606.1
NM_001324226.1 Intron NP_001311155.1
NM_001324227.1 Intron NP_001311156.1
NM_001324228.1 Intron NP_001311157.1
NM_001324229.1 Intron NP_001311158.1
NM_001324230.1 Intron NP_001311159.1
NM_004809.4 Intron NP_004800.2
XM_005254792.4 Intron XP_005254849.1
XM_006720773.3 Intron XP_006720836.1

View Full Product Details