Product Details

SNP ID
rs138585940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34752582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAGATTGACTTTCTCTTGGCCT[G/T]AGCCCCTCGCACAGCCAGCTTGATC
Phenotype
MIM: 607058
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GJD2 PubMed Links

Gene Details

Gene
GJD2
Gene Name
gap junction protein delta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020660.2 955 Missense Mutation AAG,CAG K288Q NP_065711.1
XM_017022438.1 955 Missense Mutation AAG,CAG K237Q XP_016877927.1
Gene
LOC101928174
Gene Name
uncharacterized LOC101928174
There are no transcripts associated with this gene.

View Full Product Details