Product Details

SNP ID
rs141345466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63126922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACCAGAGACAGTTATGCGAATTG[C/T]TAGCATCAGCAAAAGTCTCACCATG
Phenotype
MIM: 608440
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LACTB PubMed Links

Gene Details

Gene
LACTB
Gene Name
lactamase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288585.1 560 Missense Mutation GCT,GTT A163V NP_001275514.1
NM_032857.4 560 Missense Mutation GCT,GTT A163V NP_116246.2
NM_171846.3 560 Missense Mutation GCT,GTT A163V NP_741982.1
XM_011521180.1 560 Missense Mutation GCT,GTT A163V XP_011519482.1

View Full Product Details